2-(Trimethylsilyl)thiazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79265-30-8
:
Formula:C₆H₁₁NSSi
MDL Number:MFCD00066274
:157.31 g/mol
Boiling Pt:53…55 °C (10 mmHg)
Density:0.985
Flash Pt:57 °C (135 °F)
ADR:3,III
:GHS02
:GHS07