2-(Trimethylsilyl)benzothiazole
Catalog # SILNSILS19075.5
Supplier: CHEMPUR SILANE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:157.31 g/mol
CAS Number:32137-73-8
Formula:C₆H₁₁NSSi
MDL Number:MFCD00068185