Serotonin hydrochloride 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:153-98-0
:
Formula:C₁₀H₁₃ClN₂O
MDL Number:MFCD00012686
:212.68 g/mol
Melting Pt:168…172 °C
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Loss on drying:≤2.5 % (115°C, 3 hrs) (vacuum)
- Appearance of solution:(1% in methanol) clear to slightly hazy,