6-Fluorotryptamine
Glass bottle for solids, 1 g
Catalog # FLUO010197-1G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:575-85-9
- Formula:C₁₀H₁₁FN₂
- MDL Number:MFCD00044761
- Molecular Weight:178.21 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



