Order Entry
Germany
ContactUsLinkComponent
5-Fluorotryptamine hydrochloride, Sigma-Aldrich®
5-Fluorotryptamine hydrochloride, Sigma-Aldrich®
Catalog # SIALF7001-250MG
Supplier:  Merck
CAS Number:  
5-Fluorotryptamine hydrochloride, Sigma-Aldrich®
Catalog # SIALF7001-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :214.67 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:220-307-5
CAS Number:2711-58-2
Formula:C₁₀H₁₂ClFN₂
MDL Number:MFCD00051081