5-Fluorotryptamine hydrochloride
Catalog # FLUO010196-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.67 g/mol
EINECS:220-307-5
CAS Number:2711-58-2
Formula:C₁₀H₁₂ClFN₂
MDL Number:MFCD00051081