Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-5-methoxyindole-2-boronic acid
1-Boc-5-methoxyindole-2-boronic acid, Sigma-Aldrich®
1-Boc-5-methoxyindole-2-boronic acid, Sigma-Aldrich®
1-Boc-5-methoxyindole-2-boronic acid, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1-Boc-5-methoxyindole-2-boronic acid, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
290331-71-4
:
Formula:
C₁₄H₁₈BNO₅
MDL Number:
MFCD04039006
:
291.11 g/mol
:
M-Clarity™ quality level = MQ100