Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-ethoxybenzaldehyde
5-Bromo-2-ethoxybenzaldehyde
5-Bromo-2-ethoxybenzaldehyde
5-Bromo-2-ethoxybenzaldehyde
Supplier:
Thermo Scientific
Certificates
Expand image
5-Bromo-2-ethoxybenzaldehyde
Supplier:
Thermo Scientific
About this item
CAS Number:
79636-94-5
:
Formula:
C₉H₉BrO₂
MDL Number:
MFCD00016600
:
229.07 g/mol