Pesticide reference standard, 2,3,5-Trichlorophenol 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL17773500IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:933-78-8
- Description:2,3,5-Trichlorophenol 10 µg/ml in isooctane
- Formula:C₆H₃Cl₃O
- Description:2,3,5-Trichlorophenol