
1-Methyl-4-piperidone, Sigma-Aldrich®
Catalog # 8.20828.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:113.16 g/mol
- Boiling Point:60 - 62 °C (19 hPa)
- Flash Point:58 °C
- Refractive Index:1.4605 (20 °C, 589 nm)
- Density:0.97 g/cm3 (20 °C)
- pH:12 (100 g/l, H2O, 20 °C)
- Synonyms:N-Methyl-γ-piperidone
- Hill Formula:C6H11NO
Specification Test Results
About this item
:
:113.16 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:215-895-5
UN:1993
:GHS02
Storage Temperature:Ambient
CAS Number:1445-73-4
Formula:C₆H₁₁NO
MDL Number:MFCD00006191