Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Propanol
2-Propanol ≥70% (v/v)
2-Propanol ≥70% (v/v)
2-Propanol ≥70% (v/v)
ALCH31070/5
:
Alcoholes Montplet
:
2-Propanol ≥70% (v/v)
ALCH31070/5
:
Alcoholes Montplet
:
Pk:
5 L
Seq:
5190
:
:
60.1 g/mol
:
0.8…0.92 g/cm³ (20 °C)
:
Ambient
:
200-661-7
:
1987
:
3,II
:
GHS02
:
GHS07
:
67-63-0
:
(CH₃)₂CHOH
:
MFCD00011674