2-Propanol ≥70% (v/v), sterile
Catalog # 148-0123
Supplier: Texwipe
CAS Number:
Specifications
- Pk:4
- Seq:0004
- Pk Info:4 x 3,8 l
Specifications
About this item
:
:60.1 g/mol
Density:0.8…0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-661-7
UN:1987
ADR:3,II
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674