Carboxymethyl cellulose sodium salt, Sigma-Aldrich®
Catalog # SIAL21904-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:9004-32-4
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472