Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Agarose
Agarose, Sigma-Aldrich®
Agarose, Sigma-Aldrich®
Agarose, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Agarose, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
9012-36-6
:
MDL Number:
MFCD00081294
Melting Pt:
60…90 °C
Storage Temperature:
Ambient
EINECS:
232-731-8