Bisacrylamide, powder
Supplier: G-Biosciences
Certificates
About this item
CAS Number:110-26-9
:
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625
:154.17 g/mol
Boiling Pt:125 °C (3.33 hPa)
Melting Pt:185 °C
Density:1.235 g/cm³ (20 °C)
Flash Pt:138 °C
EINECS:203-750-9
:GHS07
Storage Temperature:Refrigerator