Flat flanges, grounded, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
These flanges are available with or without groove.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Made from clear DURAN® borosilicate glass 3.3.
Frequently Bought Together