Orders LinkContactUsLinkComponent
Evaporating dishes
Evaporating dishes
Supplier:   BOCHEM
Evaporating dishes
Supplier:  BOCHEM
Certificates
Evaporating dishes
Evaporating dishes
4 Options Available Below
Product Details & Documents
Made of nickel 99,5 %.
  • Good chemical resistance
  • Very good temperatrure resistance
  • Easy to clean
Without spout.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 45 ml
    19 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,50 €
    57,50 €
    1 * 1 ST
       57,50 €
  • 45 ml
    16 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,40 €
    66,40 €
    1 * 1 ST
       66,40 €
  • 73 ml
    19 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,20 €
    65,20 €
    1 * 1 ST
       65,20 €
  • 100 ml
    16 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,90 €
    82,90 €
    1 * 1 ST
       82,90 €