About this item
Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol
- CAS Number:100-02-7
- Formula:C₆H₅NO₃
- MDL Number:MFCD00007331
- Molecular Weight:139.11 g/mol
- Merck Index:13,06654
- Boiling Pt:279 °C (1013 hPa)
- Melting Pt:110…114 °C
- Density:1.48 g/cm³ (20 °C)
- Flash Pt:169 °C
- Storage Temperature:Ambient
- UN:1663
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- EINECS:202-811-7
2 Options Available Below
Product Details & Documents
Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol
- CAS Number:100-02-7
- Formula:C₆H₅NO₃
- MDL Number:MFCD00007331
- Molecular Weight:139.11 g/mol
- Merck Index:13,06654
- Boiling Pt:279 °C (1013 hPa)
- Melting Pt:110…114 °C
- Density:1.48 g/cm³ (20 °C)
- Flash Pt:169 °C
- Storage Temperature:Ambient
- UN:1663
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- EINECS:202-811-7
Documents
Specifications for Product Family
Specification Test Results
- Hill Formula:C₆H₅NO₃
- Chemical formula:O₂NC₆H₄OH
- Molar Mass:139.11 g/mol
- Boiling point:279 °C (1013 hPa) (decomposition)
- Density:1.48 g/cm³ (20 °C)
- Flash point:169 °C
- Ignition temperature:510 °C
- Melting Point:110 - 114 °C
- pH value:4.4 (5 g/l, H₂O, 24 °C)
- Bulk density:550 kg/m³
- Solubility:16 g/l (25 °C)
- LD 50 dermal:LD50 Rat 1024 mg/kg
- Storage class:6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
- WGK:WGK 2 water endangering
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



