4-Nitrophenol
Catalog # EHERC15590400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:139.11 g/mol
Merck Index:13,06654
Boiling Pt:279 °C (1013 hPa)
Melting Pt:110…114 °C
Density:1.48 g/cm³ (20 °C)
Flash Pt:169 °C
Storage Temperature:Ambient
EINECS:202-811-7
UN:1663
ADR:6.1,III
:GHS08
:GHS07
CAS Number:100-02-7
Formula:C₆H₅NO₃
MDL Number:MFCD00007331