VWR®, Filter Tips
All tips are made of high quality, 100% pure, virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard. All products are tested for endotoxin (pyrogen) contamination and certified to USP results. Resins are pre-tested for metal contamination. Certificates are on file by lot number for all test results.
- A wider seal ensures that tips will seal easily and consistently without leaking, on old as well as new pipettes
- Guaranteed free of detectable human DNA, DNase/RNase, ATP, endotoxins and heavy metals
- Catalog No:
- 732-0571
- 732-0540
- 732-0520
- 732-0541
- 732-0542
- 732-0543
- 732-1100
- 732-0532
- 732-0536
- 732-0559
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging