Order Entry
Germany
ContactUsLinkComponent
N-Methylaniline for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
N-Methylaniline for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Environment
N-Methylaniline for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    5.62 g/l (25 °C)
  • Melting Point:
    -57 °C
  • Molar Mass:
    107.16 g/mol
  • Boiling Point:
    196 °C (1013 hPa)
  • Vapor Pressure:
    0.5 hPa (20 °C)
  • Flash Point:
    86 °C
  • Refractive Index:
    1.5704 (20 °C, 589 nm)
  • Density:
    0.99 g/cm3 (20 °C)
  • pH:
    7.6 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    500 °C

Specification Test Results

Related Information