Media bottles, graduated, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Manufactured from borosilicate glass which provides the best chemical resistance.
- Special pour lip facilitates pouring
- Black Phenolic PTFE rubber lined cap
- Very good temperature resistance
Graduations and marking patch assists in measuring volume and identifying content. Hazardous communication label included with each case of bottles allows for content ID and safety notification. Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse.
Packaging: Packaged in partitioned carton to help prevent scratch marks on bottles.