Order Entry
Germany
ContactUsLinkComponent
2-Nitroaniline
2-Nitroaniline
Catalog # EHERC15554200
Supplier:  EHRENSTORFER
CAS Number:  
2-Nitroaniline
Catalog # EHERC15554200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :138.13 g/mol
Merck Index:12,06680
Boiling Pt:284 °C (1013 hPa)
Melting Pt:68…71 °C
Density:1.446 g/cm³ (20 °C)
Flash Pt:167 °C
Storage Temperature:Ambient
EINECS:201-855-4
UN:1661
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:88-74-4
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007687