Pestles, serrated
Supplier: KARTELL
Certificates
About this item
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.
- Head made of PTFE
- Shaft made of stainless steel
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.