4-Iodoanisole
Catalog # FLUO001304-100G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Boiling Pt:237 °C (1013 hPa)
Melting Pt:48…49 °C
Density:1 g/cm³ (20 °C)
:234.04 g/mol
Storage Temperature:Ambient
EINECS:211-798-7
CAS Number:696-62-8
Formula:C₇H₇IO
MDL Number:MFCD00001056