Order Entry
Germany
ContactUsLinkComponent
2-Iodobenzoic acid
2-Iodobenzoic acid
Catalog # MOLE31585110-100G
Supplier:  Molekula
CAS Number:  
2-Iodobenzoic acid
Catalog # MOLE31585110-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :248.02 g/mol
Merck Index:13,05052
Boiling Pt:314 °C (1013 hPa)
Melting Pt:159…160 °C
Density:2.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-850-7
CAS Number:88-67-5
Formula:C₇H₅IO₂
MDL Number:MFCD00002419