


2,4,5-Trifluorophenol ≥94%
Catalog # B20395.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:148.08 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:39…42 °C
Flash Pt:55 °C (131 °F)
Storage Temperature:Ambient
UN:2921
ADR:8,II
:GHS06
:GHS05
:GHS02
CAS Number:2268-16-8
Formula:C₆H₃F₃O
MDL Number:MFCD00061215