2,3,5-Trifluorophenol
1 g
Catalog # FLUO001834-1G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:2268-15-7
- Formula:C₆H₃F₃O
- MDL Number:MFCD00075416
- Molecular Weight:148.08 g/mol
- UN:1325
- ADR:4.1,II
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



