Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Fluorophenol 99%

4-Fluorophenol 99%

Irritant
4-Fluorophenol 99%
Certificates
4-Fluorophenol 99%

2 Options Available Below

Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:44…49 °C
  • Density:1,22 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2811
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to beige to pink Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    44 to 49 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,60 €
    36,60 €
    1 * 25 g
       36,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 98,60 €
    98,60 €
    1 * 100 g
       98,60 €