Order Entry
Germany
ContactUsLinkComponent
2-Fluoro-4-nitrophenol
2-Fluoro-4-nitrophenol
Catalog # MOLE43304392-5G
Supplier:  Molekula
CAS Number:  
2-Fluoro-4-nitrophenol
Catalog # MOLE43304392-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :157.1 g/mol
Melting Pt:118…118.5 °C
CAS Number:403-19-0
Formula:C₆H₄FNO₃
MDL Number:MFCD00051970
Melting Pt:118…118,5 °C