Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-4-nitrophenol
2-Fluoro-4-nitrophenol ≥98%
2-Fluoro-4-nitrophenol ≥98%
:
Thermo Fisher Scientific
2-Fluoro-4-nitrophenol ≥98%
:
Thermo Fisher Scientific
:
403-19-0
:
:
C₆H₄FNO₃
:
MFCD00051970
:
157.1 g/mol
:
117…121 °C
:
GHS06
:
GHS07
:
Ambient
msds_ALFAA13134
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years