3,5-Difluorophenol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 2713-34-0 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 70°(158°F) Odour: Phenol-like Boiling Point: 65-68°/1mm Beilstein Reference: 2078616 UN No.: 3261 MDL No.: MFCD00002255
- CAS Number:2713-34-0
- Formula:C₆H₄F₂O
- MDL Number:MFCD00002255
- Molecular Weight:130.09 g/mol
- Boiling Pt:65…68 °C (1 mmHg)
- Melting Pt:51…55 °C
- Density:1.566 g/cm³ (20 °C)
- Flash Pt:70 °C (158 °F)
- Storage Temperature:Ambient
- UN:3261
- ADR:8,II
- :Density: 1.566 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 2713-34-0 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 70°(158°F) Odour: Phenol-like Boiling Point: 65-68°/1mm Beilstein Reference: 2078616 UN No.: 3261 MDL No.: MFCD00002255
- CAS Number:2713-34-0
- Formula:C₆H₄F₂O
- MDL Number:MFCD00002255
- Molecular Weight:130.09 g/mol
- Boiling Pt:65…68 °C (1 mmHg)
- Melting Pt:51…55 °C
- Density:1.566 g/cm³ (20 °C)
- Flash Pt:70 °C (158 °F)
- Storage Temperature:Ambient
- UN:3261
- ADR:8,II
- :Density: 1.566 g/cm³ (20 °C)
Dokumente
Certificates
Search by Lot Numbermsds_ALFAA15177
Sicherheitsdatenblatt
Organic Building Blocks and Intermediates



