2,4-Difluorophenol ≥98%
About this item
Purity: 98+% CAS No.: 367-27-1 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Refractive Index: 1.4860 Odour: Phenol-like Melting Point: 21-24° Boiling Point: 52-54°/14mm Beilstein Reference: 1934803 UN No.: 2921 MDL No.: MFCD00009715
- CAS Number:367-27-1
- Formula:C₆H₄F₂O
- MDL Number:MFCD00009715
- Molecular Weight:130.09 g/mol
- Boiling Pt:52…54 °C (14 mmHg)
- Melting Pt:21…24 °C
- Density:1.362
- Flash Pt:56 °C (132 °F)
- Storage Temperature:Ambient
- EINECS:206-688-0
- UN:2921
- ADR:8,II
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 367-27-1 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Refractive Index: 1.4860 Odour: Phenol-like Melting Point: 21-24° Boiling Point: 52-54°/14mm Beilstein Reference: 1934803 UN No.: 2921 MDL No.: MFCD00009715
- CAS Number:367-27-1
- Formula:C₆H₄F₂O
- MDL Number:MFCD00009715
- Molecular Weight:130.09 g/mol
- Boiling Pt:52…54 °C (14 mmHg)
- Melting Pt:21…24 °C
- Density:1.362
- Flash Pt:56 °C (132 °F)
- Storage Temperature:Ambient
- EINECS:206-688-0
- UN:2921
- ADR:8,II
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



