4-Fluoroanisole
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:459-60-9
:
Formula:C₇H₇FO
MDL Number:MFCD00000348
:126.13 g/mol
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–43.5 °C
Density:1.1178 g/cm³ (18.3 °C)
EINECS:207-295-7
UN:3271
ADR:3,III