Order Entry
Germany
ContactUsLinkComponent
4-Fluoroanisole 99%
4-Fluoroanisole 99%
Catalog # ACRO119310250
CAS Number:  
undefined
4-Fluoroanisole 99%
Catalog # ACRO119310250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4867 to 1.4887 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :126.13 g/mol
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–45 °C
Density:1.11 g/cm³
Flash Pt:43 °C
EINECS:207-295-7
UN:1993
ADR:3,III
 :GHS02
CAS Number:459-60-9
Formula:C₇H₇FO
MDL Number:MFCD00000348