Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-p-anisaldehyde
3-Fluoro-p-anisaldehyde
3-Fluoro-p-anisaldehyde
3-Fluoro-p-anisaldehyde
Supplier:
FLUOROCHEM
Certificates
Expand image
3-Fluoro-p-anisaldehyde
Supplier:
FLUOROCHEM
About this item
CAS Number:
351-54-2
:
Formula:
C₈H₇FO₂
MDL Number:
MFCD00003349
:
154.14 g/mol
EINECS:
206-514-3