2,6-Difluorobenzaldehyde
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:437-81-0
:
Formula:C₇H₄F₂O
MDL Number:MFCD00010293
Boiling Pt:194 °C (1013 hPa)
Melting Pt:15…17 °C
Density:1,31 g/cm³ (20 °C)
:142.1 g/mol
Storage Temperature:Ambient