Fluorobenzene
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:462-06-6
:
Formula:C₆H₅F
MDL Number:MFCD00000280
:96.1 g/mol
Merck Index:13,04197
Boiling Pt:85.1 °C (1013 hPa)
Melting Pt:–41 °C
Density:1.023 g/cm³ (20 °C)
Flash Pt:–15 °C
Storage Temperature:Ambient
EINECS:207-321-7
UN:2387
ADR:3,II