Fluorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:462-06-6
:
Formula:C₆H₅F
MDL Number:MFCD00000280
:96.1 g/mol
Merck Index:14,4170
Boiling Pt:84…85 °C
Melting Pt:–42 °C
Density:1.025
Flash Pt:–15 °C (5 °F)
Storage Temperature:Ambient
EINECS:207-321-7
UN:2387
ADR:3,II
:GHS02
:GHS07