Fluorobenzene 99%
:462-06-6
:
:C₆H₅F
:MFCD00000280
:96.1 g/mol
:13,04197
:85 °C (760 mmHg)
:–42 °C
:1.02 g/cm³
:–15 °C
:Ambient
:207-321-7
:2387
:3,II
:GHS02
:GHS05
:GHS09
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4643 to 1.4663 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %