Base moulds, stainless steel
Supplier: Simport Scientific
Certificates
About this item
Made of stainless steel. Sizes fit most cassettes. Superior thermal exchange due to rounded corners. These moulds are for all applications in specimen embedding with Simport embedding rings and cassettes. Manufactured from high quality stainless steel for optimal thermal conductivity.
- The moulds have a highly polished surface for easy paraffin block removal
- Well corners are rounded for optimal paraffin ribboning