Orders LinkContactUsLinkComponent
Safety shields
Safety shields
242×365 mm
Catalog # KART571
Supplier:   KARTELL
Safety shields
Catalog # KART571
Supplier:  KARTELL
Safety shields
Safety shields
242×365 mm
Price
261,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PMMA protection shield, transparent, thickness: 5 mm, provides laboratory and healthcare personnel with facial protection from hazardous substances, aerosol and splashing.
  • Free standing
  • Porable and stable
  • Fitted with non skid feet
Specifications
  • W×H:
    242×365 mm