Order Entry
Germany
Orders LinkContactUsLinkComponent
Goggles, Cobra

Goggles, Cobra

Anti-UV, anti-scratch, anti-fog, Clear PC, Transparent/black
Catalog # 111-0670
Supplier:   Bollé Safety
Goggles, Cobra
Catalog # 111-0670
Supplier:  Bollé Safety
Goggles, Cobra
Goggles, Cobra

Anti-UV, anti-scratch, anti-fog, Clear PC, Transparent/black

Price
18,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Low profile goggles with soft foam seal, adjustable strap and non slip bridge. This version of the Cobra combines the comfort of an eyeshield with the fit and seal of an all-purpose goggle.

  • Clear anti-static PC lenses for 180° panoramic field of view
  • Interchangeable comfort foam
  • Lenses with anti-fog and anti-scratch coating for improved resistance

Weight: 26 g

Certifications: EN 166, EN 170

Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    Transparent/black
  • Lenses:
    Clear PC
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.