Orders LinkContactUsLinkComponent
Bottle Carriers
Bottle Carriers
Supplier:   KARTELL
Bottle Carriers
Supplier:  KARTELL
Certificates
Bottle Carriers
2 Options Available Below
Product Details & Documents
PP, coloured.
  • Ergonomic design of the handle
  • Can be easily stacked
  • Very good chemical and temperature resistance
Designed to carry 6 and 4 bottles, with maximum diameter up to 90 mm and 110 mm respectively.
Product Family Options
Product Information
  • Colour
    For
    Container max. Ø
    Availability
    Price
  • KART395
    Bottle carriers
    Bottle carriers
    Red
    Up to six bottles
    95 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 131,00 €
    131,00 €
    1 * 5 ST
       131,00 €
  • KART396
    Bottle carriers
    Bottle carriers
    Yellow
    Up to four bottles
    120 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 152,00 €
    152,00 €
    1 * 5 ST
       152,00 €