


2,4,6-Trichlorophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-06-2
:
Formula:C₆H₃Cl₃O
MDL Number:MFCD00002172
:197.45 g/mol
Merck Index:12,09773
Boiling Pt:244.5 °C (1013 hPa)
Melting Pt:∼66 °C
Density:1.675 g/cm³ (25 °C)
EINECS:201-795-9
UN:2020
ADR:6.1,III
:M-Clarity™ quality level = MQ100
Flash Pt:99 °C
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient