Pesticide reference standard, 2,4,6-Trichlorophenol 1000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA17774600ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:88-06-2
- Description:2,4,6-Trichlorophenol 1,000 µg/ml in methanol
- Formula:C₆H₃Cl₃O
- Description:2,4,6-Trichlorophenol