

2,4,5-Trichlorophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-95-4
:
Formula:C₆H₃Cl₃O
MDL Number:MFCD00002170
:197.45 g/mol
Merck Index:12,09772
Boiling Pt:253 °C (1013 hPa)
Melting Pt:68 °C
Density:1.68 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:202-467-8
:GHS07
:GHS09
Storage Temperature:Ambient