Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Chlorophenol ≥99%

Irritant
Environment
4-Chlorophenol ≥99%
Certificates
4-Chlorophenol ≥99%
4-Chlorophenol ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 106-48-9 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 115°(239°F) Odour: Phenol-like Boiling Point: 217-220° Beilstein Reference: 507004 Merck Reference: 14,2154 UN No.: 2020 MDL No.: MFCD00002318

Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:14,2154
  • Boiling Pt:217…220 °C
  • Melting Pt:39…43 °C
  • Density:1.306
  • Flash Pt:115 °C (239 °F)
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III
Product Family Options

Product Information