

3-Chlorophenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-43-0
:
Formula:C₆H₅ClO
MDL Number:MFCD00002256
Merck Index:13,02173
Boiling Pt:∼214 °C (1013 hPa)
Melting Pt:31…34 °C
Density:1.245 g/cm³ (20 °C)
Flash Pt:97 °C
EINECS:203-582-6
UN:2020
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:128.56 g/mol
:GHS07
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:27.7 g/l (20 °C)
- Melting Point:31 - 34 °C
- Molar Mass:128.55 g/mol
- Boiling Point:216.8 °C (1013 hPa)
- Vapor Pressure:1 hPa (44 °C)
- Flash Point:97 °C
- Density:1.27 g/cm3 (45 °C)
- pH:3.5 (27.7 g/l, H2O, 20 °C)
- Ignition Point:600 °C
- Hill Formula:C6H5ClO
- Chemical Formula:3-(Cl)C6H4OH