Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
Supplier:   KARTELL
Microplates, 96-well
Supplier:  KARTELL
Certificates
Microplates, 96-well
6 Options Available Below
Product Details & Documents
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
  • Aseptic or sterile
  • Single-wrapped
Ordering information: PS covers are sold separately.
Product Family Options
Product Information
  • Colour
    Volume
    No. of wells
    Availability
    Price
  • KART2620
    96-well plate, flat bottom, aseptic
    96-well plate, flat bottom, aseptic
    Clear
    370 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 114,00 €
    114,00 €
    1 * 50 ST
       114,00 €
  • KART2621
    96-well plate, U-bottom, aseptic
    96-well plate, U-bottom, aseptic
    Clear
    367 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 113,00 €
    113,00 €
    1 * 50 ST
       113,00 €
  • KART2622
    96-well plates, V-bottom, aseptic
    96-well plates, V-bottom, aseptic
    Clear
    288 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 113,00 €
    113,00 €
    1 * 50 ST
       113,00 €
  • KART2620/S
    96-well plate, flat bottom, sterile
    96-well plate, flat bottom, sterile
    Clear
    370 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 120,00 €
    120,00 €
    1 * 50 ST
       120,00 €
  • KART2621/S
    96-well plate, U-bottom, sterile
    96-well plate, U-bottom, sterile
    Clear
    367 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 120,00 €
    120,00 €
    1 * 50 ST
       120,00 €
  • KART2622/S
    96-well plate, V-bottom, sterile
    96-well plate, V-bottom, sterile
    Clear
    288 µl
    96
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 120,00 €
    120,00 €
    1 * 50 ST
       120,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lids for 96 well microtitre plates, PS, aseptic
    Lids for 96 well microtitre plates, PS, aseptic
    KART2623
    Log in for availability
    1 * 50 ST - 93,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids for 96 well microtitre plates, PS, sterile
    Lids for 96 well microtitre plates, PS, sterile
    KART2623/S
    Log in for availability
    1 * 50 ST - 97,30 €